Learn More
HLA-B7 Monoclonal Antibody (BB7.1), APC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
145.00€ - 348.00€
Spécification
Antigène | HLA-B7 |
---|---|
Clone | BB7.1 |
Concentration | 5 μl/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16360714
|
Affymetrix eBioscience
17-5917-41 |
25 Tests |
145.00€
25 pièces |
Expédition estimée: 14-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16370714
|
Affymetrix eBioscience
17-5917-42 |
100 Tests |
348.00€
100 pièces |
Expédition estimée: 14-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Description: This BB7.1 monoclonal antibody recognizes the human major histocompatibility complex MHC class I molecule HLA-B7, which is a member of the HLA-B family. MHC class I exists as a heterodimer composed of a specific alpha chain associated with a beta 2-microglobulin. MHC class I molecules are expressed on all nucleated cells. Applications Reported: This BB7.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This BB7.1 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser
HLA-B7 is one of the polymorphic proteins of human MHC Class I found on all nucleated cells. This surface protein is a heterodimer consisting of the specific transmembrane HLA-B chain paired with Beta-2 microglobulin. Classical MHC Class I molecules play a central role in adaptive immunity by presenting to T cells antigenic peptides derived either from the cell itself (classical presentation) or extracellular antigens (cross-presentation). The ligands of MHC class I include T cell receptor (TCR) and a costimulatory molecule CD8. Hundreds of HLA-B alleles have been described. Typing for these polymorphisms is routinely done for transplantation.Spécification
HLA-B7 | |
5 μl/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
MHC class I antigen B*7, HLA class I histocompatibility antigen B-7 alpha chain | |
HLA-B | |
IgG1 κ | |
Affinity Chromatography | |
Antibody |
BB7.1 | |
Flow Cytometry | |
APC | |
Mouse | |
Human | |
P01889 | |
3106 | |
Papain solubilized HLA-B7 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
HLA-B |