missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-G Monoclonal Antibody (87G), FITC, Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA110365
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody recognizes both membrane-bound and soluble forms of HLA-G (HLA-G1 and HLA-G5). It will not cross-react with mouse or rat.
HLA-G belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. HLA-G is expressed on fetal derived placental cells. The heavy chain is approximately 45 kDa and its gene contains 8 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domain, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exon 6 encodes the cytoplasmic tail. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.Spécification
HLA-G | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P17693 | |
HLA-G | |
HLA-B27 transgenic mice were imunized with H-2 identical murine cells transfected with and expressing genes encoding HLA-G and human beta2-microglobulin. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry | |
87G | |
FITC | |
HLA-G | |
acrosomal protein SP-10; Acrosomal vesicle protein 1; ACRV1; b2 microglobulin; D11S4365; DADB-15K14.8; HLA class I histocompatibility antigen, alpha chain G; HLA class I molecule; HLA G antigen; HLA-6.0; HLAG; HLA-G; HLA-G histocompatibility antigen, class I, G; LOC768350; major histocompatibility complex, class I, G; MHC class I antigen G; MHC-G; MHCIY; MHC-like class I Y; sHLA-G; Soluble HLA class I histocompatibility antigen, alpha chain G; SP-10; SPACA2; sperm protein 10 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
3135 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |