missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-G (soluble) Monoclonal Antibody (2A12), FITC, Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA110364
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody does not cross-react with the full-length HLA-G1 isoform and therefore is able to distinguish between secreted HLA-G5 and HLA-G6 isoforms from shedded HLA-G1.
HLA-G belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. HLA-G is expressed on fetal derived placental cells. The heavy chain is approximately 45 kDa and its gene contains 8 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domain, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exon 6 encodes the cytoplasmic tail. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.Spécification
HLA-G (soluble) | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P17693 | |
HLA-G | |
C-terminal amino acid sequence (22-mer) of soluble HLA-G5 and HLA-G6 proteins coupled to ovalbumin. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
2A12 | |
FITC | |
HLA-G | |
acrosomal protein SP-10; Acrosomal vesicle protein 1; ACRV1; b2 microglobulin; D11S4365; DADB-15K14.8; HLA class I histocompatibility antigen, alpha chain G; HLA class I molecule; HLA G antigen; HLA-6.0; HLAG; HLA-G; HLA-G histocompatibility antigen, class I, G; LOC768350; major histocompatibility complex, class I, G; MHC class I antigen G; MHC-G; MHCIY; MHC-like class I Y; sHLA-G; Soluble HLA class I histocompatibility antigen, alpha chain G; SP-10; SPACA2; sperm protein 10 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
3135 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |