Learn More
HMBS Mouse anti-Human, Unconjugated, Clone: 1G12H3, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 67456-1-IG-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The HMBS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HMBS. This antibody recognizes human antigen. The HMBS antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
HMBS | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, ELISA | |
1G12H3 | |
Unconjugated | |
150 μL | |
3145 | |
IgG1 |