missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMG20B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14582-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Mutations of the human BRCA2 gene confer susceptibility to breast cancer. A structural DNA binding component has been identified and is designated as BRCA2-Associated Factor 35 (BRAF35). There is a close association of the BRAF35/BRCA2 complex with condensed chromatin coincident with histone H3 phosphorylation. Experimental evidence suggests a role for the BRCA2/BRAF35 complex in the modulation of cell cycle progression.Spécification
HMG20B | |
Polyclonal | |
Unconjugated | |
HMG20B | |
BRAF25, BRAF35, BRCA2 associated factor 35, high mobility group 20B, HMG box containing protein 20B, HMG20B, HMGX2, HMGXB2, PP7706, pp8857, SMARCE1 related protein, SMARCE1r, SOXL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10362, 15353, 362825 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9P0W2, Q9Z104 | |
Hmg20b | |
HMG20B Fusion Protein Ag6103 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |