missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOIP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16289-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Lyn proto oncogene (Lcl/Yes related novel protein tyrosine kinase) is encoded by the Lyn gene located on chromosome 8 in humans and belongs to the Src family kinase. It is a non-receptor tyrosine kinase that acts as a mediator in transducing cellular signaling. The most well studied expression of Lyn is in hematopoietic cell types, both lymphoid and myeloid cells with exclusion in T lymphocytes. It has also been observed that Lyn plays diverse roles such as in regulation of B cell receptor signaling, mast cell signaling and Epithelial - Mesenchymal Transition (EMT).Spécification
HOIP | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q924T7, Q96EP0 | |
HOIL 1 interacting protein, HOIP, ring finger protein 31, RNF31, ZIBRA | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
268749, 364386, 55072 |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
RNF31 | |
Liquid | |
Rnf31 | |
HOIP Fusion Protein Ag9440 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Mouse, Rat, Human |