missing translation for 'onlineSavingsMsg'
Learn More

HOXA2 Rabbit anti-Human, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 25044-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16803585

  • 112.71€ / 20µL

Expédition estimée
02-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.
Spécification

Spécification

HOXA2
0.36 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O43364, P31246
homeobox A2, Homeobox protein Hox 1K, Homeobox protein Hox A2, HOX1K, HOXA2
Rabbit
IgG
20 μL
-20°C
Primary
103690123, 3199
Western Blot
Unconjugated
HOXA2
Liquid
HOXA2
HOXA2 Fusion Protein Ag19224
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Rat
Vidéos
FDS
Documents