missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD17B7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16925-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Responsible for the reduction of the keto group on the C-3 of sterols.Spécification
HSD17B7 | |
Polyclonal | |
Unconjugated | |
HSD17B7P2 | |
HSD17B7P2 | |
HSD17B7 Fusion Protein Ag10315 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Hsd17b_2, HSD17B7, HSD17B7P2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
158160 | |
-20°C | |
Liquid |