missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPA4 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21206-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody well recognized the endogenous HSPA4 protein in mouse brain/ testis tissues.
The HSP 70 family is composed of four highly conserved proteins: HSP 70, HSC 70, GRP 75 and GRP 78. HSP 70 expression is strongly induced in response to heat stress. HSP 70 and HSC 70 play key roles in the cytosolic endoplasmic reticulum and mitochondrial import machinery and are found in both the cytosol and nucleus of mammalian cells. GRP 78 is localized in the endoplasmic reticulum, where it receives imported secretory proteins and is involved in the folding and translocation of nascent peptide chains. GRP 75 expression is restricted to the mitochondrial matrix and aids in the translocation and folding of nascent polypeptide chains of both nuclear and mitochondrial origin. It has been postulated that members of the HSP 70 family act as force-generating motors, relying on the hydrolysis of ATP for their activitySpécification
HSPA4 | |
Polyclonal | |
Unconjugated | |
HSPA4 | |
APG 2, APG2, Heat shock 70 kDa protein 4, heat shock 70kDa protein 4, HS24/P52, HSP70.4, hsp70RY, HSPA4, RY | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15525, 266759, 3308 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O88600, P34932, Q61316 | |
HSPA4 | |
HSPA4 Fusion Protein Ag15581 | |
150 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |