Learn More
HTR5A Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 28221-1-AP-20UL
Informations supplémentaires : Poids : 0.00020kg
Description
The HTR5A antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human HTR5A. This antibody recognizes human, mouse antigen. The HTR5A antibody has been validated for the following applications: IHC,ELISA analysis.
Spécification
HTR5A | |
Polyclonal | |
Unconjugated | |
20 μL | |
3361 | |
Liquid |
Immunohistochemistry, ELISA | |
550 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |