missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human 4-1BB Ligand / TNFSF9 Protein, Fc Tag
sur devis
Spécification
Numéro d’adhésion | NP_003802 |
---|---|
Poids moléculaire | 47.5 kDa |
Source | Human 4-1 BB Ligand, Fc Tag (41 L-H5257) is expressed from human 293 cells (HEK293). It contains AA Ala 50 - Glu 254 (Accession # NP_003802). |
Immunogène | Ala 50 - Glu 254 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15875867
|
ACROBiosystems
41L-H5257-50UG |
50 µg |
Cet article est actuellement indisponible ou discontinu.
Veuillez consulter la page produit pour de possible alternative dans l’onglet |
||||||
Description
Tumor necrosis factor ligand superfamily member 9 (4-1BBL) is also known as 4-1BB ligand, CD137L or TNFSF9, which is a cytokine that binds to TNFRSF9. 4-1BBL is the high affinity ligand of 4-1BB. 4-1BBL induces the proliferation of activated peripheral blood T-cells. Also, 4-1BBL may have a role in activation-induced cell death (AICD). Furthermore, 4-1BBL may play a role in cognate interactions between T-cells and B-cells/macrophages. As for diseases, 4-1BBL is involved in cancers, infectious diseases and autoimmune diseases.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 47.5 kDa. The protein migrates as 52 kDa under reducing (R) condition (SDS-PAGE).Spécification
NP_003802 | |
Human 4-1 BB Ligand, Fc Tag (41 L-H5257) is expressed from human 293 cells (HEK293). It contains AA Ala 50 - Glu 254 (Accession # NP_003802). | |
-20°C | |
4-1 BB Ligand | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |
47.5 kDa | |
Ala 50 - Glu 254 | |
4-1 BB Ligand,TNFSF9,CD137L | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |