missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human 4-1BB Receptor Recombinant Protein, PeproTech®

E. coli Recombinant Protein

Marque:  Gibco™ 310-15-1MG

Informations supplémentaires : Poids : 0.16500kg

 Afficher plus de versions de ce produit

Code produit. 17892573

  • 2884.00€ / 1mg
Expédition estimée: 25-10-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

CD137, also known as TNFRSF9 or 4-1BB, is an inducible costimulatory molecule expressed mainly on activated T cells. Its ligand, known as 4-1BBL, is expressed on activated macrophages, mature B cells, hematopoietic stem cells, and myeloid progenitor cells. CD137 signaling leads to maintaining the survival of activated T cells and CD8+ memory T cells, and clonal expansion of T cells, but also to suppressing myelopoiesis and dendritic cell development. Triggered CD137 induces a cytokine release profile regulating peripheral monocyte survival. Soluble forms of CD137 may provide negative control mechanism for some immune responses.
Spécification

Spécification

Q07011
protein with no preservative
17.7
1 mg
-20°C
<1 EU/µg
TNFRSF9
Unconjugated
MERTRSLQDP CSNCPAGTFC DNNRNQICSP CPPNSFSSAG GQRTCDICRQ CKGVFRTRKE CSSTSNAECD CTPGFHCLGA GCSMCEQDCK QGQELTKKGC KNCCFGTFND QKRGICRPWT NCSLDGKSVL VNGTKERDVV CGPSPADLSP GASSVTPPAP AREPGHS
≥ 98% by SDS-PAGE gel and HPLC analyses.
ELISA, Functional Assay, Western Blot
3604
Human 4-1 BB Receptor
E. coli
RUO
4 1 BB; 41 BB; 4-1 BB; 4-1 BB ligand receptor; A930040I11Rik; AA408498; AI325004; Cd137; CD137 antigen; CDw137; EGK_00183; homolog of mouse 4-1 BB; ILA; induced by lymphocyte activation (ILA); interleukin-activated receptor, homolog of mouse Ly63; Ly63; receptor protein 4-1 BB; s4 1 BB; s4 1 BB; s41BB; sCD137; secreted CD137 antigen; soluble 4 1 BB; soluble 4 1 BB; soluble 41 BB; soluble CD137; T cell antigen ILA; T-cell antigen 4-1 BB; T-cell antigen 4-1 BB homolog; T-cell antigen ILA; TNF receptor superfamily member 9; TNFRSF9; Tumor necrosis factor receptor superfamily member 9; tumor necrosis factor receptor superfamily, member 9
Human
Recombinant
Lyophilized
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions