Learn More
Abnova™ Human AFF1 Partial ORF (NP_005926.1, 71 a.a. - 170 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00004299-Q01.10ug
Informations supplémentaires : Poids : 0.00010kg
Description
DISCONTINUED: The record for PBM1 has been withdrawn by HGNC.
Sequence: LSTKSHTHRLDASENRLGKPKYPLIPDKGSSIPSSSFHTSVHHQSIHTPASGPLSVGNISHNPKMAQPRTEPMPSLHAKSCGPPDSQHLTQDRLGQEGFGSpécification
NP_005926.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LSTKSHTHRLDASENRLGKPKYPLIPDKGSSIPSSSFHTSVHHQSIHTPASGPLSVGNISHNPKMAQPRTEPMPSLHAKSCGPPDSQHLTQDRLGQEGFG | |
RUO | |
AFF1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4299 | |
AFF1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AF-4/AF4/AF4-MLL/MGC134969/MLL/AF4/MLLT2/PBM1 | |
AFF1 | |
Recombinant | |
wheat germ expression system |