Learn More
ACROBiosystems Human AIMP1 / EMAP2 / SCYE1 Protein, His Tag
362.89€ - 2201.57€
Spécification
Numéro d’adhésion | AAH14051.1 |
---|---|
Poids moléculaire | 35.2 kDa |
Source | Human AIMP1, His Tag (EM2-H5142) is expressed from E.coli cells. It contains AA Ala 2 - Lys 312 (Accession # Q12904-1). |
Immunogène | Ala 2 - Lys 312 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15876637
|
ACROBiosystems
EM2-H5142-100UG |
100 μg |
362.89€
100µg |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15886637
|
ACROBiosystems
EM2-H5142-1MG |
1 mg |
2201.57€
1mg |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Aminoacyl tRNA synthase complex-interacting multifunctional protein 1 (AIMP1) is also known as multisynthase complex auxiliary component p43 and endothelial monocyte-activating polypeptide II (EMAP-II). AIMP1 is a cytokine that may be induced by apoptosis and is also released from professional antigen-presenting cells such as dendritic cells. The release of AIMP1 renders the tumor-associated vasculature sensitive to tumor necrosis factor. Furthermore, AIMP1 binds tRNA and stimulates the catalytic activity of cytoplasmic arginyl-tRNA synthase. This protein possesses inflammatory cytokine activity. Also, AIMP1 negatively regulates TGF-beta signaling through stabilization of SMURF2 by binding to SMURF2 and inhibiting its SMAD7-mediated degradation.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 35.2 kDa. The protein migrates as 36 kDa under reducing (R) condition (SDS-PAGE).Spécification
AAH14051.1 | |
Human AIMP1, His Tag (EM2-H5142) is expressed from E.coli cells. It contains AA Ala 2 - Lys 312 (Accession # Q12904-1). | |
-20°C | |
AIMP1 | |
Human | |
PBS, pH7.4 | |
E.coli | |
Cytokines and Growth Factors |
35.2 kDa | |
Ala 2 - Lys 312 | |
AIMP1,EMAP2,EMAPII,HLD3,SCYE1,p43,EMAP-2,EMAP-II,AIMP-1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |