missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AKAP5 Full-length ORF (NP_004848.2, 1 a.a. - 427 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marque:  Abnova™ H00009495-P01.10ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16123742

  • 335.00€ / 10µg
Expédition estimée: 28-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein binds to the RII-beta regulatory subunit of PKA, and also to protein kinase C and the phosphatase calcineurin. It is predominantly expressed in cerebral cortex and may anchor the PKA protein at postsynaptic densities (PSD) and be involved in the regulation of postsynaptic events. It is also expressed in T lymphocytes and may function to inhibit interleukin-2 transcription by disrupting calcineurin-dependent dephosphorylation of NFAT. [provided by RefSeq]

Sequence: METTISEIHVENKDEKRSAEGSPGAERQKEKASMLCFKRRKKAAKALKPKAGSEAADVARKCPQEAGASDQPEPTRGAWASLKRLVTRRKRSESSKQQKPLEGEMQPAINAEDADLSKKKAKSRLKIPCIKFPRGPKRSNHSKIIEDSDCSIKVQEEAEILDIQTQTPLNDQATKAKSTQDLSEGISRKDGDEVCESNVSNSITSGEKVISVELGLDNGHSAIQTGTLILEEIETIKEKQDVQPQQASPLETSETDHQQPVLSDVPPLPAIPDQQIVEEASNSTLESAPNGKDYESTEIVAEETKPKDTELSQESDFKENGITEEKSKSEESKRMEPIAIIITDTEISEFDVTKSKNVPKQFLISAENEQVGVFANDNGFEDRTSEQYETLLIETASSLVKNAIQLSIEQLVNEMASDDNKINNLLQ
Spécification

Spécification

NP_004848.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
73.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
AKAP5
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
9495
AKAP5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AKAP75/AKAP79/H21
AKAP5
Recombinant
wheat germ expression system
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions