Learn More
Abnova™ Human AKAP9 Partial ORF (NP_671700, 3812 a.a. - 3911 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00010142-Q01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. Alternate splicing of this gene results in at least two isoforms that localize to the centrosome and the Golgi apparatus, and interact with numerous signaling proteins from multiple signal transduction pathways. These signaling proteins include type II protein kinase A, serine/threonine kinase protein kinase N, protein phosphatase 1, protein phosphatase 2a, protein kinase C-epsilon and phosphodiesterase 4D3. [provided by RefSeq]
Sequence: EKTDSFYHSSGGLELYGEPRHTTYRSRSDLDYIRSPLPFQNRYPGTPADFNPGSLACSQLQNYDPDRALTDYITRLEALQRRLGTIQSGSTTQFHAGMRRSpécification
NP_671700 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EKTDSFYHSSGGLELYGEPRHTTYRSRSDLDYIRSPLPFQNRYPGTPADFNPGSLACSQLQNYDPDRALTDYITRLEALQRRLGTIQSGSTTQFHAGMRR | |
RUO | |
AKAP9 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10142 | |
AKAP9 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AKAP350/AKAP450/CG-NAP/HYPERION/KIAA0803/MU-RMS-40.16A/PRKA9/YOTIAO | |
AKAP9 | |
Recombinant | |
wheat germ expression system |