missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AP1M2 Full-length ORF (NP_005489.2, 1 a.a. - 423 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marque:  Abnova™ H00010053-P01.10ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16124422

  • 335.00€ / 10µg
Expédition estimée: 10-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a subunit of the heterotetrameric adaptor-related protein comlex 1 (AP-1), which belongs to the adaptor complexes medium subunits family. This protein is capable of interacting with tyrosine-based sorting signals. [provided by RefSeq]

Sequence: MSASAVFILDVKGKPLISRNYKGDVAMSKIEHFMPLLVQREEEGALAPLLSHGQVHFLWIKHSNLYLVATTSKNANASLVYSFLYKTIEVFCEYFKELEEESIRDNFVIVYELLDELMDFGFPQTTDSKILQEYITQQSNKLETGKSRVPPTVTNAVSWRSEGIKYKKNEVFIDVIESVNLLVNANGSVLLSEIVGTIKLKVFLSGMPELRLGLNDRVLFELTGRSKNKSVELEDVKFHQCVRLSRFDNDRTISFIPPDGDFELMSYRLSTQVKPLIWIESVIEKFSHSRVEIMVKAKGQFKKQSVANGVEISVPVPSDADSPRFKTSVGSAKYVPERNVVIWSIKSFPGGKEYLMRAHFGLPSVEKEEVEGRPPIGVKFEIPYFTVSGIQVRYMKIIEKSGYQALPWVRYITQSGDYQLRTS
Spécification

Spécification

NP_005489.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
74.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
AP1M2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
10053
AP1M2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AP1-mu2/HSMU1B/MU-1B/MU1B/mu2
AP1M2
Recombinant
wheat germ expression system
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions