Learn More
Abnova™ Human ATG4A Full-length ORF (NP_443168.2, 1 a.a. - 398 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Spécification
Numéro d’adhésion | NP_443168.2 |
---|---|
À utiliser avec (application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formule | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Identification génétique (Entrez) | 115201 |
Poids moléculaire | 71.8kDa |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16107203
|
Abnova™
H00115201-P01.25UG |
25 ug |
508.00€
25µg |
Expédition estimée: 30-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16197193
|
Abnova™
H00115201-P01.10UG |
10 ug |
335.00€
10µg |
Expédition estimée: 30-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Autophagy is the process by which endogenous proteins and damaged organelles are destroyed intracellularly. Autophagy is postulated to be essential for cell homeostasis and cell remodeling during differentiation, metamorphosis, non-apoptotic cell death, and aging. Reduced levels of autophagy have been described in some malignant tumors, and a role for autophagy in controlling the unregulated cell growth linked to cancer has been proposed. This gene encodes a member of the autophagin protein family. The encoded protein is also designated as a member of the C-54 family of cysteine proteases. Transcript variants that encode distinct isoforms have been identified. [provided by RefSeq]
Sequence: MESVLSKYEDQITIFTDYLEEYPDTDELVWILGKQHLLKTEKSKLLSDISARLWFTYRRKFSPIGGTGPSSDAGWGCMLRCGQMMLAQALICRHLGRDWSWEKQKEQPKEYQRILQCFLDRKDCCYSIHQMAQMGVGEGKSIGEWFGPNTVAQVLKKLALFDEWNSLAVYVSMDNTVVIEDIKKMCRVLPLSADTAGDRPPDSLTASNQSKGTSAYCSAWKPLLLIVPLRLGINQINPVYVDAFKECFKMPQSLGALGGKPNNAYYFIGFLGDELIFLDPHTTQTFVDTEENGTVNDQTFHCLQSPQRMNILNLDPSVALGFFCKEEKDFDNWCSLVQKEILKENLRMFELVQKHPSHWPPFVPPAKPEVTTTGAEFIDSTEQLEEFDLEEDFEILSVSpécification
NP_443168.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
71.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
APG4A/AUTL2 | |
ATG4A | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
115201 | |
ATG4A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
ATG4A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |