Learn More
ACROBiosystems Human Azurocidin / CAP37 / AZU1 Protein, His Tag
Marque: ACROBiosystems AZ1-H5225-100UG
Informations supplémentaires : Poids : 0.00001kg
Description
Azurocidin (AZU1) is also known as Heparin-binding protein (HBP), Cationic antimicrobial protein CAP37, is a neutrophil granule-derived antibacterial and monocyte- and fibroblast-specific chemotactic glycoprotein. which belongs to the peptidase S1 family and elastase subfamily. AZU1 / HBP contains 1 peptidase S1 domain. AZU1 binds heparin. The cytotoxic action of AZU1 is limited to many species of Gram-negative bacteria; this specificity may be explained by a strong affinity of the very basic N-terminal half for the negatively charged lipopolysaccharides that are unique to the Gram-negative bacterial outer envelope. AZU1 may play a role in mediating recruitment of monocytes in the second wave of inflammation.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.0 kDa. The protein migrates as 34-42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
AAH69495 | |
100 μg | |
Ile 27 - Pro 250 | |
AZU1,Azurocidin,HBP,AZAMP,AZU,CAP37,HUMAZUR,NAZC | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
25.0 kDa | |
Human Azurocidin, His Tag (AZ1-H5225) is expressed from human 293 cells (HEK293). It contains AA Ile 27 - Pro 250 (Accession # AAH69495). | |
-20°C | |
Azurocidin | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |