missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human Bach2 Full-Length Orf (Aai66613.1, 1 A.A.-841 A.A.) Recombinant Protein With Gst Tag At N-Terminal.

Recombinant proteins expressed from in vitro wheat germ system. Such proteins mirror the conformation and folding of the native proteins in the eukaryotic biological system.

Marque:  Abnova™ H00060468-P01.25ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16123213

  • 467.00€ / 25µg

Expédition estimée
11-01-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

  • Good Folding
  • Excellent amino acids labeling

Immunogen Sequence: MSVDEKPDSPMYVYESTVHCTNILLGLNDQRKKDILCDVTLIVERKEFRAHRAVLAACSEYFWQALVGQTKNDLVVSLPEEVTARGFGPLLQFAYTAKLLLSRENIREVIRCAEFLRMHNLEDSCFSFLQTQLLNSEDGLFVCRKD

Spécification

Spécification

AAI66613.1
Liquid
60468
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
BACH2
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer.
BACH2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BACH2
GST
Vidéos
FDS
Documents

For Research Use Only