missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BAPX1 Partial ORF (NP_001180, 53 a.a. - 111 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marque:  Abnova™ H00000579-Q01.25ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16152131

  • 545.00€ / 25µg
Expédition estimée: 12-07-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a member of the NK family of homeobox-containing proteins. The encoded protein may play a role in skeletal development. [provided by RefSeq]

Sequence: RLFGERDAGALGGAEDSLLASPAGTRTAAGRTAESPEGWDSDSALSEENESRRRCADAR
Spécification

Spécification

NP_001180
Liquid
579
BAPX1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BAPX1/MGC138171/NKX3.2/NKX3B
NKX3-2
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.23kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RLFGERDAGALGGAEDSLLASPAGTRTAAGRTAESPEGWDSDSALSEENESRRRCADAR
RUO
NKX3-2
Wheat Germ (in vitro)
GST
Vidéos
FDS
Documents
Certificats
Promotions

Promotions