missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human BCMA / TNFRSF17 Protein,His Tag DMF Filed
HPLC-verified
471.77€ - 3229.78€
Spécification
Numéro d’adhésion | NP_001183.2 |
---|---|
Poids moléculaire | 7.8 kDa |
Source | Human BCMA Protein, His Tag (HPLC-verified) (BCA-H522y) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 54 (Accession # Q02223-1). |
Immunogène | Met 1 - Ala 54 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15876027
|
ACROBiosystems
BCA-H522Y-100UG |
100 μg |
471.77€
100µg |
Expédition estimée: 13-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15886027
|
ACROBiosystems
BCA-H522Y-1MG |
1 mg |
3229.78€
1mg |
Expédition estimée: 30-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Tumor necrosis factor receptor superfamily member 17 (TNFRSF17) is also known as B-cell maturation protein (BCMA), CD antigen CD269, which is a member of the TNF-receptor superfamily. TNFRSF17 contains one TNFR-Cys repeat. TNFRSF17 is expressed in mature B-cells, but not in T-cells or monocytes. TNFRSF17 is receptor for TNFSF13B/BLyS/BAFF and TNFSF13/APRIL. TNFRSF17 promotes B-cell survival and plays a role in the regulation of humoral immunity. TNFRSF17 can activate NF-kappa-B and JNK.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 7.8 kDa. The protein migrates as 12 kDa and 14-18 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_001183.2 | |
Human BCMA Protein, His Tag (HPLC-verified) (BCA-H522y) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 54 (Accession # Q02223-1). | |
-20°C | |
BCMA | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
7.8 kDa | |
Met 1 - Ala 54 | |
TNFRSF17,CD269,BCM,BCMA | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |