missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human BTLA Protein, His Tag

Marque:  ACROBiosystems BTA-H52E0-50UG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15876067

  • 353.08€ / 50µg
Date d’expédition estimée: 29-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

B- and T-lymphocyte attenuator (BTLA) is also known as B- and T-lymphocyte-associated protein, CD antigen CD272. BTLA contains one Ig-like V-type (immunoglobulin-like) domain. As a lymphocyte inhibitory receptor, BTLA / CD272 inhibits lymphocytes during immune response. BTLA / CD272 can interact with tyrosine phosphatases PTPN6/SHP-1 and PTPN11/SHP-2, and interact with TNFRSF14/HVEM.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.6 kDa. The protein migrates as 25-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification

Spécification

AAP44003.1
HEK293
Human
15.6 kDa
50 µg
Lys 31 - Ser 150
BTLA,CD272
Unconjugated
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
Immune Checkpoint Proteins
Powder
>90% as determined by SDS-PAGE
Human BTLA, His Tag (BTA-H52E0) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Ser 150 (Accession # AAP44003.1).
-20°C
BTLA
His Tag
Vidéos
FDS
Documents
Certificats
Promotions

Promotions