Learn More
ACROBiosystems Human C-Reactive Protein (CRP) Protein, His Tag
181.45€ - 641.12€
Spécification
Numéro d’adhésion | NP_000558.2 |
---|---|
Poids moléculaire | 23.9 kDa |
Source | Human C-Reactive Protein, His Tag (CRP-H5226) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 224 (Accession # NP_000558.2). |
Immunogène | Gln 19 - Pro 224 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15866477
|
ACROBiosystems
CRP-H5226-250UG |
250 μg |
181.45€
250µg |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15856477
|
ACROBiosystems
CRP-H5226-1MG |
1 mg |
641.12€
1mg |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
C-reactive protein (CRP) is a member of the pentraxin family of proteins that are characterized by a cyclic pentameric structure. Human CRP gene encodes a 224 amino acids precursor. The mature human CRP protein has 206 amino acids that are noncovalently linked to form the pentameter. Human CRP shares 71% and 64% amino acid sequence homology with mouse and rat respectively. CRP, synthesized by hepatocytes, is a major acute phase serum protein in human. IL6, IL1 and glucocorticoids are the major inducer of the CRP gene. The physiological role of CRP is to bind to phosphocholine expressed on the surface of dead or dying cells (and some types of bacteria) in order to activate the complement system. CRP binds to phosphocholine on microbes and damaged cells and enhances phagocytosis by macrophages. Thus, CRP participates in the clearance of necrotic and apoptotic cells. CRP rises up to 50,000-fold in acute inflammation, such as infection. It rises above normal limits within 6 hours, and peaks at 48 hours. Its half-life is constant, and therefore its level is mainly determined by the rate of production. It has been shown that high levels of CRP in humans is associated with an increased risk of cardiovascular diseases.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.9 kDa. The protein migrates as 26 kDa under reducing (R) condition (SDS-PAGE).Spécification
NP_000558.2 | |
Human C-Reactive Protein, His Tag (CRP-H5226) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 224 (Accession # NP_000558.2). | |
-20°C | |
C-Reactive Protein | |
Human | |
Tris-NaCl buffer pH8.0 | |
HEK293 | |
Bio-Markers and CD Antigens |
23.9 kDa | |
Gln 19 - Pro 224 | |
CRP,C-reactive Protein | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |