missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human C12orf51 Partial ORF (XP_497354, 3105 a.a. - 3209 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00283450-Q01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
Sequence: DQHIEFFWGALEMFTQEELCKFIKFACNQERIPFTCPCKDGGPDTAHVPPYPMKIAPPDGTAGSPDSRYIRVETCMFMIKLPQYSSLEIMLEKLRCAIHYREDPLSpécification
XP_497354 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.29kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DQHIEFFWGALEMFTQEELCKFIKFACNQERIPFTCPCKDGGPDTAHVPPYPMKIAPPDGTAGSPDSRYIRVETCMFMIKLPQYSSLEIMLEKLRCAIHYREDPL | |
RUO | |
C12orf51 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
283450 | |
C12orf51 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp586O1022/FLJ10510/FLJ30092/FLJ30208/FLJ34154/KIAA0614/MGC126531 | |
C12orf51 | |
Recombinant | |
wheat germ expression system |