Learn More
ACROBiosystems Human C1q R1 / CD93 Protein, His Tag
350.80€ - 2419.31€
Spécification
Numéro d’adhésion | AAH28075 |
---|---|
Poids moléculaire | 59.2 kDa |
Source | Human C1q R1, His Tag (C11-H5228) is expressed from human 293 cells (HEK293). It contains AA Thr 22 - Lys 580 (Accession # AAH28075). |
Immunogène | Thr 22 - Lys 580 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15866077
|
ACROBiosystems
C11-H5228-1MG |
1 mg |
2419.31€
1mg |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15876077
|
ACROBiosystems
C11-H5228-50UG |
50 μg |
350.80€
50µg |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
CD antigen CD93 is also known as Complement component C1q receptor (C1QR1). which belongs to the Group XIV C-Type lectin family, a group containing two other members, endosialin (CD248) and thrombomodulin. CD93 contains one C-type lectin domain and five EGF-like domains. CD93 is highly expressed in endothelial cells, platelets, cells of myeloid origin, such as monocytes and neutrophils. CD93 is receptor for C1q, mannose-binding lectin (MBL2) and pulmonary surfactant protein A (SPA). CD93 may mediate the enhancement of phagocytosis in monocytes and macrophages upon interaction with soluble defense collagens and may play a role in intercellular adhesion.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.2 kDa. The protein migrates as 70-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
AAH28075 | |
Human C1q R1, His Tag (C11-H5228) is expressed from human 293 cells (HEK293). It contains AA Thr 22 - Lys 580 (Accession # AAH28075). | |
-20°C | |
C1q R1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
59.2 kDa | |
Thr 22 - Lys 580 | |
CD93,CDw93,C1QR1,MXRA4,C1qR,C1qR(P),C1qRP,ECSM3 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |