missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CAMKK2 Partial ORF (AAH26060, 1 a.a. - 130 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marque:  Abnova™ H00010645-Q01.25ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16125502

  • 467.00€ / 25µg

Expédition estimée
22-12-2020

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Sequence: MSSCVSSQPSSNRAAPQDELGGRGSSSSESQKPCEALRGLSSLSIHLGMESFIVVTECEPGCAVDLGLARDRPLEADGQEVPLDSSGSQARPHLSGRKLSLQERSQGGLAAGGSLDMNGRCICPSLPYSP
Spécification

Spécification

AAH26060
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
39.93kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MSSCVSSQPSSNRAAPQDELGGRGSSSSESQKPCEALRGLSSLSIHLGMESFIVVTECEPGCAVDLGLARDRPLEADGQEVPLDSSGSQARPHLSGRKLSLQERSQGGLAAGGSLDMNGRCICPSLPYSP
RUO
CAMKK2
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
10645
CAMKK2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CAMKK|CAMKKB|KIAA0787|MGC15254
CAMKK2
Yes
Vidéos
FDS
Documents