Learn More
Abnova™ Human CD302 Full-length ORF (AAH20646.1, 1 a.a. - 170 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00009936-P01.10ug
Informations supplémentaires : Poids : 0.00010kg
Description
CD302 is a C-type lectin receptor involved in cell adhesion and migration, as well as endocytosis and phagocytosis (Kato et al., 2007 [PubMed 17947679]).[supplied by OMIM]
Sequence: MISIHNEEENAFILDTLKKQWKGPDDILLGMFYDTDDASFKWFDNSNMTFDKWTDQDDDEDLVDTCAFLHIKTGEWKKGNCEVSSVEGTLCKTAIPYKRKYLSDNHILISALVIASTVILTVLGAIIWFLYKKHSDSRFTTVFSTAPQSPYNEDCVLVVGEENEYPVQFDSpécification
AAH20646.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
45.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BIMLEC/CLEC13A/DCL-1/DCL1/KIAA0022 | |
CD302 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9936 | |
CD302 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MISIHNEEENAFILDTLKKQWKGPDDILLGMFYDTDDASFKWFDNSNMTFDKWTDQDDDEDLVDTCAFLHIKTGEWKKGNCEVSSVEGTLCKTAIPYKRKYLSDNHILISALVIASTVILTVLGAIIWFLYKKHSDSRFTTVFSTAPQSPYNEDCVLVVGEENEYPVQFD | |
RUO | |
CD302 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |