missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human CD48 / SLAMF2 Protein, Fc Tag

Marque:  ACROBiosystems BC1-H5255-1MG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15876017

  • 1926.79€ / 1mg
Date d’expédition estimée: 03-01-2022

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

CD48 antigen is also known as Signaling lymphocytic activation molecule 2, SLAM family member 2 (SLAMF2), Leukocyte antigen MEM-102, BCM1 surface antigen, B-lymphocyte activation marker BLAST-1. CD48 is a 65 kDa GPI-linked protein which belongs to the CD2 subfamily of immunoglobulin superfamily molecules. CD48 antigen contains two Ig-like C2-type (immunoglobulin-like) domains and two Ig-like V-type (immunoglobulin-like) domain. CD48 interacts with CD244 in an heterophilic manner. CD48 is ligand for CD2. CD48 might facilitate interaction between activated lymphocytes. CD48 is probably involved in regulating T-cell activation.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 49.0 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification

Spécification

NP_001769.2
HEK293
Human
49.0 kDa
1 mg
Gln 27 - Ser 220
CD48,BCM1,SLAMF2,BLAST,BLAST1,MEM-102,TCT0.1,BCM-1,SLAMF-2,BLAST-1
Unconjugated
Tris with Glycine, Arginine and NaCl, pH7.5
Less than 1.0 EU / µg by the LAL method.
Immune Checkpoint Proteins
Powder
>95% as determined by reduced SDS-PAGE
Human CD48, Fc Tag (BC1-H5255) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Ser 220 (Accession # P09326-1).
-20°C
CD48
Fc Tag
Vidéos
FDS
Documents
Certificats
Promotions

Promotions