Learn More
ACROBiosystems Human CD84 / SLAMF5 Protein, His Tag
Marque: ACROBiosystems CD4-H5229-1MG
Informations supplémentaires : Poids : 0.00001kg
Description
Leukocyte differentiation antigen CD84 is also known as SLAM family member 5 (SLAMF5), which belongs to immunoglobulin (Ig) superfamily. CD84 / SLAMF5 contains one Ig-like C2-type (immunoglobulin-like) domain. CD84 plays a role as adhesion receptor functioning by homophilic interactions and by clustering. CD84 / SLAMF5 increases proliferative responses of activated T-cells and SH2D1A/SAP does not seen be required for this process. Homophilic interactions enhance interferon gamma/IFNG secretion in lymphocytes and induce platelet stimulation via a SH2D1A/SAP-dependent pathway.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.6 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
AAH20063 | |
23.6 kDa | |
1 mg | |
Lys 22 - Gly 225 | |
CD84,SLAMF5,LY9B,SLAMF5 | |
Unconjugated | |
PBS, pH7.4 | |
HEK293 | |
Human |
Powder | |
>95% as determined by SDS-PAGE | |
Human CD84, His Tag (CD4-H5229) is expressed from human 293 cells (HEK293). It contains AA Lys 22 - Gly 225 (Accession # AAH20063). | |
-20°C | |
CD84 | |
His Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Immune Checkpoint Proteins |