missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CDIPT Full-length ORF (AAH01444, 1 a.a. - 213 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marque:  Abnova™ H00010423-P01.10ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16125062

  • 302.00€ / 10µg

Expédition estimée
21-12-2020

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Sequence: MPDENIFLFVPNLIGYARIVFAIISFYFMPCCPLTASSFYLLSGLLDAFDGHAARALNQGTRFGAMLDMLTDRCSTMCLLVNLALLYPGATLFFQISMSLDVASHWLHLHSSVVRGSESHKMIDLSGNPVLRIYYTSRPALFTLCAGNELFYCLLYLFHFSEGPLVGSVGLFRMGLWVTAPIALLKSLISVIHLITAARNMAALDAADRAKKK
Spécification

Spécification

AAH01444
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
49.17kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC1328|PIS|PIS1
CDIPT
Yes
wheat germ expression system
Liquid
10423
CDIPT (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPDENIFLFVPNLIGYARIVFAIISFYFMPCCPLTASSFYLLSGLLDAFDGHAARALNQGTRFGAMLDMLTDRCSTMCLLVNLALLYPGATLFFQISMSLDVASHWLHLHSSVVRGSESHKMIDLSGNPVLRIYYTSRPALFTLCAGNELFYCLLYLFHFSEGPLVGSVGLFRMGLWVTAPIALLKSLISVIHLITAARNMAALDAADRAKKK
RUO
CDIPT
Wheat Germ (in vitro)
GST
Vidéos
FDS
Documents