missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human CLEC3B / Tetranectin Protein, His Tag, low endotoxin

low endotoxin

Marque:  ACROBiosystems CLB-H5226-100UG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15866447

  • 332.90€ / 100µg
Date d’expédition estimée: 28-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

C-type lectin domain family 3 member B (CLEC3B) is also known as Tetranectin, Plasminogen kringle 4-binding protein, is a secreted homotrimer. CLEC3B contains 1 C-type lectin domain and mainly found in plasma. Tetranectin / CLEC3B binds to plasminogen and to isolated kringle 4. CLEC3B may be involved in the packaging of molecules destined for exocytosis. Decreased plasma Tetranectin or increased Tetranectin in stroma of cancers correlates with cancer progression and adverse prognosis.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.1 kDa. The protein migrates as 22 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification

Spécification

AAH11024
HEK293
Human
21.1 kDa
100 µg
Glu 22 - Val 202
CLEC3B,Tetranectin,TN,TNA
Unconjugated
PBS, pH7.4
Less than 0.1 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>95% as determined by SDS-PAGE
Human CLEC3B, His Tag, low endotoxin (CLB-H5226) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Val 202 (Accession # AAH11024).
-20°C
CLEC3B
His Tag
Vidéos
FDS
Documents
Certificats
Promotions

Promotions