missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DCUN1D4 Full-length ORF (ENSP00000263922, 1 a.a. - 197 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00023142-P01.10ug
Informations supplémentaires : Poids : 0.00010kg
Description
Sequence: MPPRKKRRPASGDDLSAKKSRHDSMYRKYDSTRIKTEEEAFSSKRCLEWFYEYAGTDDVVGPEGMEKFCEDIGVEPENVVMLVLAWKLDAQNMGYFTLQEWLKGMTSLQCDTTEKLRNTLDYLRSFLNDSTNFKLIYRYAFDFARQSKYKVINKDQWCNVLEFSRTINLDLSNYDEDGAWPVLLDEFVEWYKDKQMSSpécification
ENSP00000263922 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
49.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ42355/KIAA0276 | |
DCUN1D4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
23142 | |
DCUN1D4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPPRKKRRPASGDDLSAKKSRHDSMYRKYDSTRIKTEEEAFSSKRCLEWFYEYAGTDDVVGPEGMEKFCEDIGVEPENVVMLVLAWKLDAQNMGYFTLQEWLKGMTSLQCDTTEKLRNTLDYLRSFLNDSTNFKLIYRYAFDFARQSKYKVINKDQWCNVLEFSRTINLDLSNYDEDGAWPVLLDEFVEWYKDKQMS | |
RUO | |
DCUN1D4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |