missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human Dkk-1 Protein, Strep Tag

Marque:  ACROBiosystems DK1-H5286-25UG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15866567

  • 262.28€ / 25µg
Date d’expédition estimée: 28-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Members of the dickkopf-related protein family (DKK-1, -2, -3, and -4) are secreted proteins with two cysteine-rich domains separated by a linker region. And DKK1 takes part in embryonic development through its inhibition of the WNT signaling pathway, binds to LRP6 with high affinity and prevents the Frizzled-Wnt-LRP6 complex formation in response to Wnts. DKK1 promotes LRP6 internalization and degradation when it forms a ternary complex with the cell surface receptor Kremen.DKK1 not olny functions as a head inducer during development, but also regulates joint remodeling and bone formation, which suggests roles for DKK1 in the pathogenesis of rheumatoid arthritis and multiple myeloma. More recently research reported, DKK1 impacts eye development from a defined developmental time point on, and is critical for lens separation from the surface ectoderm via β-catenin mediated Pdgfrα and E-cadherin expression.

This protein carries a twin strep tag at the C-terminus. The protein has a calculated MW of 28.8 kDa. The protein migrates as 37-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification

Spécification

NP_036374.1
HEK293
Human
28.8 kDa
25 µg
Thr 32 - His 266
DKK1,SK
Unconjugated
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>95% as determined by SDS-PAGE
Human Dkk-1, Strep Tag (DK1-H5286) is expressed from human 293 cells (HEK293). It contains AA Thr 32 - His 266 (Accession # NP_036374.1).
-20°C
Dkk-1
Strep Tag
Vidéos
FDS
Documents
Certificats
Promotions

Promotions