missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DUSP21 Full-length ORF (NP_071359.2, 1 a.a. - 190 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marque:  Abnova™ H00063904-P01.10ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16123293

  • 310.00€ / 10µg

Expédition estimée
21-12-2020

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Sequence: MTASASSFSSSQGVQQPSIYSFSQITRSLFLSNGVAANDKLLLSSNRITAIVNASVEVVNVFFEGIQYIKVPVTDARDSRLYDFFDPIADLIHTIDMRQGRTLLHCMAGVSRSASLCLAYLMKYHSMSLLDAHTWTKSRRPIIRPNNGFWEQLINYEFKLFNNNTVRMINSPVGNIPDIYEKDLRTMISM
Spécification

Spécification

NP_071359.2
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
47.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LMWDSP21|MGC149878
DUSP21
Yes
wheat germ expression system
Liquid
63904
DUSP21 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTASASSFSSSQGVQQPSIYSFSQITRSLFLSNGVAANDKLLLSSNRITAIVNASVEVVNVFFEGIQYIKVPVTDARDSRLYDFFDPIADLIHTIDMRQGRTLLHCMAGVSRSASLCLAYLMKYHSMSLLDAHTWTKSRRPIIRPNNGFWEQLINYEFKLFNNNTVRMINSPVGNIPDIYEKDLRTMISM
RUO
DUSP21
Wheat Germ (in vitro)
GST
Vidéos
FDS
Documents