missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DZIP1 Partial ORF (NP_945319, 341 a.a. - 450 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00022873-Q01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
Sequence: GTLKDAHEFKEDRSPYPQDFHNVMQLLDSQESKWTARVQAIHQEHKKEKGRLLSHIEKLRTSMIDDLNASNVFYKKRIEELGQRLQEQNELIITQRQQIKDFTCNPLNSISpécification
NP_945319 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GTLKDAHEFKEDRSPYPQDFHNVMQLLDSQESKWTARVQAIHQEHKKEKGRLLSHIEKLRTSMIDDLNASNVFYKKRIEELGQRLQEQNELIITQRQQIKDFTCNPLNSI | |
RUO | |
DZIP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
22873 | |
DZIP1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DZIP/DZIPt1/KIAA0996/RP11-23E3.3 | |
DZIP1 | |
Recombinant | |
wheat germ expression system |