Learn More
Abnova™ Human EDA2R Full-length ORF (NP_068555.1, 1 a.a. - 297 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00060401-P01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
EDA-A1 and EDA-A2 are two isoforms of ectodysplasin that are encoded by the anhidrotic ectodermal dysplasia (EDA) gene. Mutations in EDA give rise to a clinical syndrome characterized by loss of hair, sweat glands, and teeth. The protein encoded by this gene specifically binds to EDA-A2 isoform. This protein is a type III transmembrane protein of the TNFR (tumor necrosis factor receptor) superfamily, and contains 3 cysteine-rich repeats and a single transmembrane domain but lacks an N-terminal signal peptide. Multiple alternatively spliced transcript variants have been found for this gene, but some variants lack sufficient support. [provided by RefSeq]
Sequence: MDCQENEYWDQWGRCVTCQRCGPGQELSKDCGYGEGGDAYCTACPPRRYKSSWGHHRCQSCITCAVINRVQKVNCTATSNAVCGDCLPRFYRKTRIGGLQDQECIPCTKQTPTSEVQCAFQLSLVEADAPTVPPQEATLVALVSSLLVVFTLAFLGLFFLYCKQFFNRHCQRGGLLQFEADKTAKEESLFPVPPSKETSAESQVSENIFQTQPLNPILEDDCSSTSGFPTQESFTMASCTSESHSHWVHSPIECTELDLQKFSSSASYTGAETLGGNTVESTGDRLELNVPFEVPSPSpécification
NP_068555.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
59.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EDA-A2R/EDAA2R/TNFRSF27/XEDAR | |
EDA2R | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
60401 | |
EDA2R (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDCQENEYWDQWGRCVTCQRCGPGQELSKDCGYGEGGDAYCTACPPRRYKSSWGHHRCQSCITCAVINRVQKVNCTATSNAVCGDCLPRFYRKTRIGGLQDQECIPCTKQTPTSEVQCAFQLSLVEADAPTVPPQEATLVALVSSLLVVFTLAFLGLFFLYCKQFFNRHCQRGGLLQFEADKTAKEESLFPVPPSKETSAESQVSENIFQTQPLNPILEDDCSSTSGFPTQESFTMASCTSESHSHWVHSPIECTELDLQKFSSSASYTGAETLGGNTVESTGDRLELNVPFEVPSP | |
RUO | |
EDA2R | |
Wheat Germ (in vitro) | |
GST | |
Liquid |