missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human EIF4B Full-length ORF (AAH98437.1, 1 a.a. - 611 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marque:  Abnova™ H00001975-P01.10ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16124431

  • 335.00€ / 10µg
Expédition estimée: 27-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Sequence: MAASAKKKNKKGKTISLTDFLAEDGGTGGGSTYVSKPVSWADETDDLEGDVSTTWHSNDDDVYRAPPIDRSILPTAPRAAREPNIDRSRLPKSPPYTAFLGNLPYDVTEESIKEFFRGLNISAVRLPREPSNPERLKGFGYAEFEDLDSLLSALSLNEESLGNRRIRVDVADQAQDKDRDDRSFGRDRNRDSDKTDTDWRARPATDSFDDYPPRRGDDSFGDKYRDRYDSDRYRDGYRDGYRDGPCRDMDRYGGRDRYDDRGSRDYDRGYDSRIGSGRRAFGSGYRRDDDYRGGGDRYEDRYDRRDDRSWSSRDDYSRDDYRRDDRGPPQRPKLNLKPRSTPKEDDSSASTSQSTRAASIFGGAKPVDTAAREREVEERLQKEQEKLQRQLDEPKLERRPRERHPSWRSEETQERERSRTGSESSQTGTSTTSSRNARRRESEKSLENETLNKEEDCHSPTSKPPKPDQPLKVMPAPPPKENAWVKRSSNPPARSQSSDTEQQSPTSGGGKVAPAQPSEEGPGRKDENKVDGMNAPKGQTGNSSRGPGDGGNRDHWKESDRKDGKKDQDSRSAPEPKKPEENPASKFSSASKYAALSVDGEDENEGEDYAE
Spécification

Spécification

AAH98437.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
95.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EIF-4B/PRO1843
EIF4B
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
1975
EIF4B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAASAKKKNKKGKTISLTDFLAEDGGTGGGSTYVSKPVSWADETDDLEGDVSTTWHSNDDDVYRAPPIDRSILPTAPRAAREPNIDRSRLPKSPPYTAFLGNLPYDVTEESIKEFFRGLNISAVRLPREPSNPERLKGFGYAEFEDLDSLLSALSLNEESLGNRRIRVDVADQAQDKDRDDRSFGRDRNRDSDKTDTDWRARPATDSFDDYPPRRGDDSFGDKYRDRYDSDRYRDGYRDGYRDGPCRDMDRYGGRDRYDDRGSRDYDRGYDSRIGSGRRAFGSGYRRDDDYRGGGDRYEDRYDRRDDRSWSSRDDYSRDDYRRDDRGPPQRPKLNLKPRSTPKEDDSSASTSQSTRAASIFGGAKPVDTAAREREVEERLQKEQEKLQRQLDEPKLERRPRERHPSWRSEETQERERSRTGSESSQTGTSTTSSRNARRRESEKSLENETLNKEEDCHSPTSKPPKPDQPLKVMPAPPPKENAWVKRSSNPPARSQSSDTEQQSPTSGGGKVAPAQPSEEGPGRKDENKVDGMNAPKGQTGNSSRGPGDGGNRDHWKESDRKDGKKDQDSRSAPEPKKPEENPASKFSSASKYAALSVDGEDENEGEDYAE
RUO
EIF4B
Wheat Germ (in vitro)
GST
Liquid
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions