Learn More
ACROBiosystems Human EMMPRIN / CD147 Protein, Fc Tag
Marque:  ACROBiosystems CD7-H5259-100UG
Informations supplémentaires : Poids : 0.00001kg
Code produit. 15876267
  • 262.28€ / 100µg
Date d’expédition estimée: 05-01-2022
Description
Description
CD147 is also known as Basigin (BSG), or extracellular matrix metalloproteinase inducer (EMMPRIN). The human basigin protein contains 269 amino acids that form two heavily glycosylated C2 type immunoglobulin-like domains at the N-terminal extracellular portion. A second form of basigin has also been characterized that contains one additional immunoglobulin-like domain in its extracellular portion. As members of the immunoglobulin superfamily play fundamental roles in intercellular recognition involved in various immunologic phenomena, differentiation, and development, basigin is thought also to play a role in intercellular recognition and regulate several distinct functions, such as spermatogenesis, expression of the monocarboxylate transporter and the responsiveness of lymphocytes. Basigin is a type I integral membrane receptor that has many ligands, including the cyclophilin (CyP) proteins Cyp-A and CyP-B and certain integrins.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 46.3 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification
Spécification
NP_940991.1
HEK293
Human
46.3 kDa
100 µg
Ala 22 - His 205
Basigin,BSG,5F7,CD147,EMMPRIN,M6,OK,TCSF
Unconjugated
50 mM Tris, 100 mM Glycine, pH7.5
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>95% as determined by SDS-PAGE
Human EMMPRIN, Fc Tag (CD7-H5259) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - His 205 (Accession # NP_940991.1).
-20°C
EMMPRIN
Fc Tag