missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human EPHA10 Partial ORF (NP_001004338, 444 a.a. - 543 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marque:  Abnova™ H00284656-Q01.25ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16122747

  • 508.00€ / 25µg
Expédition estimée: 27-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Ephrin receptors, the largest subfamily of receptor tyrosine kinases (RTKs), and their ephrin ligands are important mediators of cell-cell communication regulating cell attachment, shape, and mobility in neuronal and epithelial cells (Aasheim et al., 2005 [PubMed 15777695]). See MIM 179610 for additional background on Eph receptors and ephrins.[supplied by OMIM]

Sequence: RYKDSFAAAGYGSLEAVAEMTAQDLVSLGISLAEHREALLSGISALQARVLQLQGQGVQDNKNLSVIWCLFEWQLLDHCIAGVEGLLPPSSRTGVSRICV
Spécification

Spécification

NP_001004338
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RYKDSFAAAGYGSLEAVAEMTAQDLVSLGISLAEHREALLSGISALQARVLQLQGQGVQDNKNLSVIWCLFEWQLLDHCIAGVEGLLPPSSRTGVSRICV
RUO
EPHA10
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
284656
EPHA10 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ16103/FLJ33655/MGC43817
EPHA10
Recombinant
wheat germ expression system
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions