Learn More
ACROBiosystems Human ErbB4 / Her4 Protein, Fc Tag
Marque: ACROBiosystems ER4-H5251-1MG
Informations supplémentaires : Poids : 0.00001kg
Description
Receptor tyrosine-protein kinase erbB-4 (ErbB4) is also known as Her4, is a single-pass type I transmembrane glycoprotein that is a member of the ErbB family of tyrosine kinase receptors. ErbB family members serve as receptors for the epidermal growth factor (EGF) family of growth factors. ErbB4 is expressed in normal skeletal muscle, heart, pituitary, brain and several breast carcinomas. ERBB4 contains multiple furin-like cysteine rich domains, a tyrosine kinase domain, a phosphotidylinositol-3 kinase binding site and a PDZ domain binding motif. The protein binds to and is activated by neuregulins-2 and -3, heparin-binding EGF-like growth factor and betacellulin. Ligand binding induces a variety of cellular responses including mitogenesis and differentiation. Multiple proteolytic events allow for the release of a cytoplasmic fragment and an extracellular fragment. ErbB4 appears to play important roles in neuronal development, development of the heart and cancer. ERBB4 has been shown to interact with: DLG4, NRG1, STAT5A and YAP1. Mutations in this gene have been associated with cancer. Other single-nucleotide polymorphisms and a risk haplotype have been linked to schizophrenia.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 96.0 kDa. The protein migrates as 90-106 kDa and 116-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_005226.1 | |
1 mg | |
Gln 26 - Pro 651 | |
HER4,ErbB4,MGC138404,p180erbB4 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
96.0 kDa | |
Human ErbB4, Fc Tag (ER4-H5251) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Pro 651 (Accession # Q15303-1). | |
-20°C | |
ErbB4 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |