missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human ErbB4 / Her4 Protein, Fc Tag

311.92€ - 1919.45€

Spécification

Numéro d’adhésion NP_005226.1
Poids moléculaire 96.0 kDa
Source Human ErbB4, Fc Tag (ER4-H5251) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Pro 651 (Accession # Q15303-1).
Immunogène Gln 26 - Pro 651
Conditions de stockage -20°C
Consulter d'autres spécifications

Produits 2
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
15866687
Afficher les documents Détails de la promotion
ACROBiosystems
ER4-H5251-100UG
100 μg
311.92€
100µg
Voir les produits de remplacement
Économisez jusqu'à 
»
Expédition estimée: 10-04-2024
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
15876687
Afficher les documents Détails de la promotion
ACROBiosystems
ER4-H5251-1MG
1 mg
1919.45€
1mg
Voir les produits de remplacement
Économisez jusqu'à 
»
Expédition estimée: 10-04-2024
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description

Description

Receptor tyrosine-protein kinase erbB-4 (ErbB4) is also known as Her4, is a single-pass type I transmembrane glycoprotein that is a member of the ErbB family of tyrosine kinase receptors. ErbB family members serve as receptors for the epidermal growth factor (EGF) family of growth factors. ErbB4 is expressed in normal skeletal muscle, heart, pituitary, brain and several breast carcinomas. ERBB4 contains multiple furin-like cysteine rich domains, a tyrosine kinase domain, a phosphotidylinositol-3 kinase binding site and a PDZ domain binding motif. The protein binds to and is activated by neuregulins-2 and -3, heparin-binding EGF-like growth factor and betacellulin. Ligand binding induces a variety of cellular responses including mitogenesis and differentiation. Multiple proteolytic events allow for the release of a cytoplasmic fragment and an extracellular fragment. ErbB4 appears to play important roles in neuronal development, development of the heart and cancer. ERBB4 has been shown to interact with: DLG4, NRG1, STAT5A and YAP1. Mutations in this gene have been associated with cancer. Other single-nucleotide polymorphisms and a risk haplotype have been linked to schizophrenia.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 96.0 kDa. The protein migrates as 90-106 kDa and 116-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification

Spécification

NP_005226.1
Human ErbB4, Fc Tag (ER4-H5251) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Pro 651 (Accession # Q15303-1).
-20°C
ErbB4
Human
PBS, pH7.4
HEK293
Bio-Markers and CD Antigens
96.0 kDa
Gln 26 - Pro 651
HER4,ErbB4,MGC138404,p180erbB4
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
Vidéos
FDS
Documents

Documents

Product Certifications