Learn More
ACROBiosystems Human Fas Ligand / TNFSF6 Protein, His Tag, active trimer
MALS verified
Marque: ACROBiosystems FAL-H5241-1MG
Informations supplémentaires : Poids : 0.00001kg
Description
Fas ligand is also known as FasL, CD178, CD95L, or TNFSF6, is a homotrimeric type-II transmembrane protein that belongs to the tumor necrosis factor (TNF) family. Its binding with its receptor induces apoptosis. Fas ligand/receptor interactions play an important role in the regulation of the immune system and the progression of cancer. Mature human Fas Ligand consists of a 179 amino acid (aa) extracellular domain (ECD), a 22 aa transmembrane segment, and a 80 aa cytoplasmic domain. Within the ECD, human Fas Ligand shares 81% and 78% aa sequence identity with mouse and rat Fas Ligand, respectively. Apoptosis triggered by Fas-Fas ligand binding plays a fundamental role in the regulation of the immune system. Its functions include:T-cell homeostasis, cytotoxic T-cell activity, immune privilege, maternal tolerance, tumor counterattack. Defective Fas-mediated apoptosis may lead to oncogenesis as well as drug resistance in existing tumors. Germline mutation of Fas is associated with autoimmune lymphoproliferative syndrome (ALPS), a childhood disorder of apoptosis.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 18.8 kDa. The protein migrates as 25-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
AAH17502.1 | |
1 mg | |
Pro 134 - Leu 281 | |
FASLG,ALPS1B,APT1LG1,CD178,CD95L,FASL,TNFSF6 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
18.8 kDa | |
Human Fas Ligand, His Tag, active trimer (FAL-H5241) is expressed from human 293 cells (HEK293). It contains AA Pro 134 - Leu 281 (Accession # AAH17502.1). | |
-20°C | |
Fas Ligand | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |