Learn More
ACROBiosystems Human Fetuin A / FETUA Protein, His Tag
302.42€ - 2020.12€
Spécification
Numéro d’adhésion | NP_001613.2 |
---|---|
Poids moléculaire | 38.2 kDa |
Source | Human Fetuin A, His Tag (FEA-H5221) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - Val 367 (Accession # P02765-1). |
Immunogène | Ala 19 - Val 367 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15876747
|
ACROBiosystems
FEA-H5221-1MG |
1 mg |
2020.12€
1mg |
Expédition estimée: 30-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15866747
|
ACROBiosystems
FEA-H5221-100UG |
100 μg |
302.42€
100µg |
Expédition estimée: 30-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Alpha-2-HS-glycoprotein (AHSG) is also known as Alpha-2-Z-globulin, Ba-alpha-2-glycoprotein, Fetuin-A (FETUA), is a glycoprotein that is composed of two subunits, the A and B chains, belongs to the cystatin family of proteases inhibitors and the fetuin class of plasma binding proteins. AHSG / Fetuin A is highly expressed in embryonic cells and adult hepatocytes, and is expressed to a lesser extent in monocytes/macrophages. Fetuins are carrier proteins like albumin. Fetuin-A / AHSG forms soluble complexes with calcium and phosphate and thus is a carrier of otherwise insoluble calcium phosphate. Thus AHSG / Fetuin A is a potent inhibitor of pathological calcification. Fetuin A / FETUA promotes phagocytosis of apoptotic cells and plays a role in bone development, calcium homeostasis and insulin sensitivity. Mice deficient in Fetuin-A show systemic calcification of soft tissues.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 38.2 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_001613.2 | |
Human Fetuin A, His Tag (FEA-H5221) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - Val 367 (Accession # P02765-1). | |
-20°C | |
Fetuin A | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |
38.2 kDa | |
Ala 19 - Val 367 | |
AHSG,FETUA,Fetuin A | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>98% as determined by SDS-PAGE |