Learn More
Abnova™ Human FIGLA Partial ORF (NP_001004311.1, 110 a.a. - 219 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00344018-Q01.10ug
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a protein that functions in postnatal oocyte-specific gene expression. The protein is a basic helix-loop-helix transcription factor that regulates multiple oocyte-specific genes, including genes involved in folliculogenesis and those that encode the zona pellucida. Mutations in this gene cause premature ovarian failure type 6. [provided by RefSeq]
Sequence: ATEYIQVLSDLLEGAKDSKKQDPDEQSYSNNTSESHTSSARQLSRNITQHISCAFGLKNEEEGPWADGGSGEPAHACRHSVMSTTEIISPTRSLDRFPEVELLSHRLPQVSpécification
NP_001004311.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ATEYIQVLSDLLEGAKDSKKQDPDEQSYSNNTSESHTSSARQLSRNITQHISCAFGLKNEEEGPWADGGSGEPAHACRHSVMSTTEIISPTRSLDRFPEVELLSHRLPQV | |
RUO | |
FIGLA | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
344018 | |
FIGLA (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
POF6/bHLHc8 | |
FIGLA | |
Recombinant | |
wheat germ expression system |