missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human Follistatin-like 1 / FSTL1 Protein, His Tag
302.42€ - 2020.12€
Spécification
Numéro d’adhésion | AAH00055 |
---|---|
Poids moléculaire | 33.8 kDa |
Source | Human Follistatin-like 1, His Tag (FS1-H5227) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Ile 308 (Accession # AAH00055). |
Immunogène | Glu 21 - Ile 308 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15876787
|
ACROBiosystems
FS1-H5227-100UG |
100 μg |
302.42€
100µg |
Expédition estimée: 29-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15886787
|
ACROBiosystems
FS1-H5227-1MG |
1 mg |
2020.12€
1mg |
Expédition estimée: 29-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Follistatin-related protein 1 (FSTL1) is also known as Follistatin-like protein 1 (FRP). FSTL1 is a secreted protein that contains two EF-hand domains, one follistatin-like domain, one Kazal-like domain and one VWFC domain. FSTL1 is overexpressed in synovial tissues from rheumatoid arthritis. Follistatin-like protein 1 / FSTL1 may modulate the action of some growth factors on cell proliferation and differentiation. FSTL1 binds heparin.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 33.8 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to differtent glycosylation.Spécification
AAH00055 | |
Human Follistatin-like 1, His Tag (FS1-H5227) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Ile 308 (Accession # AAH00055). | |
-20°C | |
Follistatin-like 1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |
33.8 kDa | |
Glu 21 - Ile 308 | |
FSTL1,FRP,FSL1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |