Learn More
ACROBiosystems Human Frizzled-4 / FZD4 Protein, Fc Tag
Marque: ACROBiosystems FZ4-H5250-1MG
Informations supplémentaires : Poids : 0.00001kg
Description
Frizzled-4 (FZD4) is also known as FzE4, CD344, which belongs to the G-protein coupled receptor Fz/Smo family. Most of frizzled receptors are coupled to the beta-catenin canonical signaling pathway, which leads to the activation of disheveled proteins, inhibition of GSK-3 kinase, nuclear accumulation of beta-catenin and activation of Wnt target genes. FZD4 contains one FZ (frizzled) domain. FZD4 may be involved in transduction and intercellular transmission of polarity information during tissue morphogenesis and/or in differentiated tissues. FZD4 interacts with MAGI3 and norrin (NDP).
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.9 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
AAI14528 | |
1 mg | |
Phe 37 - Glu 180 | |
FZD4,Frizzled-4,CD344,Fz-4,hFz4,FzE4 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
42.9 kDa | |
Human Frizzled-4, Fc Tag (FZ4-H5250) is expressed from human 293 cells (HEK293). It contains AA Phe 37 - Glu 180 (Accession # AAI14528). | |
-20°C | |
Frizzled-4 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |