missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human G-CSF R / CD114 Protein, Fc Tag

Marque:  ACROBiosystems GCR-H5250-1MG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15866817

  • 1210.55€ / 1mg
Date d’expédition estimée: 29-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Granulocyte Colony Stimulating Factor Receptor (G-CSFR) is also known as Cluster of Differentiation 114 (CD114), CSF3R and GCSF, is a cell-surface receptor for the granulocyte colony-stimulating factor (G-CSF), a cytokine that plays a critical role in the regulation of the activation, proliferation, differentiation, and survival of the neutrophilic granulocyte lineage. G-CSFR belongs to a family of cytokine receptors known as the hematopoietin receptor family. This type I membrane protein has a composite structure consisting of an immunoglobulin(Ig)-like domain, a cytokine receptor-homologous (CRH) domain and three fibronectin type I II (FNIII) domains in the extracellular region. G-CSFR is present mainly on precursor cells in the bone marrow, and, in response to stimulation by G-CSF, initiates cell proliferation and differentiation into mature neutrophilic granulocytes and macrophages. G-CSFR mediates the specific effect of GCSF through activating a variety of intracellular signaling cascades, including the Jak/Stat, PI3/Akt, Ras-Raf-MAP kinase, and Src family kinase pathways, and thus functions in defense against infection, inflammation and repair, and in the maintenance of steady state hematopoiesis. Mutations in this gene are a cause of Kostmann syndrome, also known as severe congenital neutropenia. Mutations in the intracellular part of this receptor are also associated with certain types of leukemia.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 92.7 kDa. The protein migrates as 100-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification

Spécification

NP_000751.1
HEK293
Human
92.7 kDa
1 mg
Glu 25 - Pro 621
CSF3R,CD114,GCSFR
Unconjugated
Tris with Glycine, Arginine and NaCl, pH7.5
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>95% as determined by SDS-PAGE
Human G-CSF R, Fc Tag (GCR-H5250) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Pro 621 (Accession # NP_000751.1).
-20°C
G-CSF R
Fc Tag
Vidéos
FDS
Documents
Certificats
Promotions

Promotions