Learn More
ACROBiosystems Human GITR / TNFRSF18 Protein, Mouse IgG2a Fc Tag, low endotoxin
low endotoxin
Marque: ACROBiosystems GIR-H525A-1MG
Informations supplémentaires : Poids : 0.00001kg
Description
Glucocorticoid-induced TNFR-related protein (GITR) is also known as Tumor necrosis factor receptor superfamily member 18 (TNFRSF18), activation-inducible TNFR family receptor (AITR), CD antigen CD357, which is a member of the tumor necrosis factor receptor (TNF-R) superfamily. GITR is receptor for TNFSF18, which seems to be involved in interactions between activated T-lymphocytes and endothelial cells and in the regulation of T-cell receptor-mediated cell death. GITR also mediated NF-kappa-B activation via the TRAF2/NIK pathway.
This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 41.4 kDa. The protein migrates as 46-49 kDa and 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_004186.1 | |
1 mg | |
Gln 26 - Glu 161 | |
AITR,GITR,TNFRSF18,CD357 | |
Unconjugated | |
Mouse IgG Fc Tag | |
Less than 0.1 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
41.4 kDa | |
Human GITR, Mouse IgG2a Fc Tag, low endotoxin (GIR-H525a) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Glu 161 (Accession # Q9Y5U5-1). | |
-20°C | |
GITR | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |