Learn More
Abnova™ Human HAAO Partial ORF (AAH29510.1, 97 a.a. - 196 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Spécification
Numéro d’adhésion | AAH29510.1 |
---|---|
À utiliser avec (application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formule | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Identification génétique (Entrez) | 23498 |
Poids moléculaire | 36.63kDa |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16107382
|
Abnova™
H00023498-Q01.25UG |
25 ug |
508.00€
25µg |
Expédition estimée: 29-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16197372
|
Abnova™
H00023498-Q01.10UG |
10 ug |
335.00€
10µg |
Expédition estimée: 29-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
3-Hydroxyanthranilate 3,4-dioxygenase is a monomeric cytosolic protein belonging to the family of intramolecular dioxygenases containing nonheme ferrous iron. It is widely distributed in peripheral organs, such as liver and kidney, and is also present in low amounts in the central nervous system. HAAO catalyzes the synthesis of quinolinic acid (QUIN) from 3-hydroxyanthranilic acid. QUIN is an excitotoxin whose toxicity is mediated by its ability to activate glutamate N-methyl-D-aspartate receptors. Increased cerebral levels of QUIN may participate in the pathogenesis of neurologic and inflammatory disorders. HAAO has been suggested to play a role in disorders associated with altered tissue levels of QUIN. [provided by RefSeq]
Sequence: ANTVGLVVERRRLETELDGLRYYVGDTMDVLFEKWFYCKDLGTQLAPIIQEFFSSEQYRTGKPIPDQLLKEPPFPLSTRSIMEPMSLDAWLDSHHRELQASpécification
AAH29510.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
3-HAO/HAO | |
HAAO | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
23498 | |
HAAO (Human) Recombinant Protein (Q01) | |
ANTVGLVVERRRLETELDGLRYYVGDTMDVLFEKWFYCKDLGTQLAPIIQEFFSSEQYRTGKPIPDQLLKEPPFPLSTRSIMEPMSLDAWLDSHHRELQA | |
RUO | |
HAAO | |
Wheat Germ (in vitro) | |
GST | |
Liquid |